missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 20 ppm C, Ricca Chemical

$256.07 - $256.07

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
R18442001C
View Documents
Ricca Chemical Company
R18442001C
1 L Amber Glass Bottle
Each for $256.07
Only null left
 
Description

Description

Specifications

Chemical Identifiers

877-24-7
204.222
23676735
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
C8H5KO4
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate

Specifications

Amber Glass Bottle
98.0,0.46,0.0041
100.0,0.48,0.0043
Liquid
Colorless
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
1 L
Organic Carbon Standard
877-24-7 , 7664-38-2 , 7732-18-5
20ppm C
1 mL = 0.020 mg C
C8H5KO4
100°C
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
204.222
23676735
Videos
SDS
Documents

Documents