missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 20 ppm C, Ricca Chemical

Catalog No. R18442001C
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R18442001C 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R18442001C Supplier Ricca Chemical Company Supplier No. R18442001C

Chemical Identifiers

CAS 877-24-7 , 7664-38-2 , 7732-18-5
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Packaging Amber Glass Bottle
Concentration or Composition Notes 20ppm C
CAS Min % 98.0, 0.46, 0.0041
CAS Max % 100.0, 0.48, 0.0043
Equivalence 1 mL = 0.020 mg C
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 1 L
Chemical Name or Material Organic Carbon Standard