missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organic Carbon Standard, 10 ppm C, Ricca Chemical
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| Odor | Odorless |
| Assay Percent Range | 10.00 ± 0.03ppm |
| Physical Form | Liquid |
| Color | Colorless to Yellow |
| pH | <2 |
| Melting Point | 0.0°C |
| Boiling Point | 100°C |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Solubility | Infinitely soluble in water |
| Show More |