missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 5 ppm C, Ricca Chemical

Catalog No. R18440504C
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R18440504C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R18440504C Supplier Ricca Chemical Company Supplier No. R18440504C
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7664-38-2 , 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) Mixture
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Packaging Amber Glass Bottle
Odor Odorless
Assay Percent Range 5.00 ± 0.05ppm
Physical Form Liquid
Color Colorless to Yellow
pH <2
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solubility Infinitely soluble in water
Solution Type Calibration Standard
Type Standard
Solvent or Matrix Water
Quantity 4 L
Grade Laboratory
Chemical Name or Material Organic Carbon Standard, 1mL = 0.005mg C, 5ppm C for Total Organic Carbon (TOC)
Show More Show Less