missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organic Carbon Standard, 1500 ppm C, Reagents
Organic Carbon Standard, 1500 ppm C, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CC205900500A
Specifications
| Organic Carbon Standard, 1500 ppm C | |
| 7732-18-5 | |
| Liquid | |
| <2 | |
| Water Analyzer | |
| Water Analysis | |
| Reagent |
| Poly Bottle | |
| 1.0049 | |
| Colorless | |
| 100°C | |
| Completely miscible | |
| 500 mL |