missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Opti-Plex™ Complexation Buffer DFS Item

Opti-Plex Complexation Buffer is a cGMP-manufactured, chemically defined, animal origin-free (AOF), methionine-free, biotin-free, low-protein transfection complexation buffer based on Opti-MEM I Reduced Serum Medium.

$81.25 - $81.25

Specifications

For Use With (Application) Protein Expression
Physical Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A4096801 DFS Item
View Documents
Gibco™
A4096801
100 mL
Each for $81.25
Only null left
 
Description

Description

Opti-Plex Complexation Buffer is a cGMP-manufactured, chemically defined, animal origin-free (AOF), methionine-free, biotin-free, low-protein transfection complexation buffer based on Opti-MEM I Reduced Serum Medium. It is designed for complexation of ExpiFectamine transfection reagents for both standard transfection applications and transfections to generate methionine-labeled proteins.

Opti-Plex Complexation Buffer features include:

  • Contains recombinant insulin, recombinant transferrin, and GlutaMAX Supplement
  • Does not contain methionine, biotin, or phenol red
  • A part of the ExpiFectamine293 Met (–) Transfection Kit (Cat. No. A39249)
Specifications

Specifications

Protein Expression
Liquid
Videos
SDS
Documents

Documents

For Research Use Only.