missing translation for 'onlineSavingsMsg'
Learn More

Ononin 97.0+%, TCI America™

Catalog No. O040510MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O040510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O040510MG Supplier TCI America Supplier No. O040510MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 486-62-4
Molecular Formula C22H22O9
Molecular Weight (g/mol) 430.409
MDL Number MFCD00017464
InChI Key MGJLSBDCWOSMHL-MIUGBVLSSA-N
Synonym Formononetin 7-O-beta-D-glucopyranoside
PubChem CID 442813
ChEBI CHEBI:7775
IUPAC Name 3-(4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one
SMILES COC1=CC=C(C=C1)C2=COC3=C(C2=O)C=CC(=C3)OC4C(C(C(C(O4)CO)O)O)O

Specifications

Melting Point 243°C
Quantity 10 mg
Formula Weight 430.41
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Ononin