accessibility menu, dialog, popup

UserName

CAS RN 486-62-4

IUPAC Name: 3-(4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4H-chromen-4-one
Synonyms: ononin formononetin glucoside
Molecular Weight (g/mol): 430.41
Molecular Formula: C22H22O9
InChi Key: MGJLSBDCWOSMHL-MIUGBVLSSA-N
SMILES: COC1=CC=C(C=C1)C1=COC2=CC(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)=CC=C2C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results