missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oncomine™ Tumor Mutation Load Assay, Chef-ready library preparation
Supplier: Thermo Fisher Scientific A37910
Spécifications
| Oncomine | |
| • 1 Oncomine Tumor Mutation Load Panel, 32 reactions, store at -5 to -30°C. • 4 cartridges Ion AmpliSeq Chef Reagents DL8, store at -5 to -30°C. • 4 cartridges Ion AmpliSeq Chef Solutions DL8, store at 2–8°C. • 4 boxes Ion AmpliSeq Supplies, store at room temperature. • 1 IonCode in PCR Plates, store at room temperature. |
|
| Targeted Sequencing Library | |
| 32 Samples | |
| Approved for shipment at Room Temperature or on Wet or Dry Ice | |
| Library Generation |
| DNA | |
| Ion GeneStudio S5 System, Ion S5™ XL System | |
| Tumor Mutation Load Assay | |
| Genome & DNA Sequencing | |
| 32 Samples |