missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oncomine™ Myeloid Research Assay
$16,620.00
Specifications
| Content And Storage | Store in freezer (-5 to -30°C). |
|---|---|
| Sample Type | DNA |
| Sequencing Type | Genome and DNA Sequencing |
| For Use With (Equipment) | Ion PGM™ System |
| Libraries | Targeted Sequencing Library |
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
A36940
|
N/A | 24 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Specifications
| Store in freezer (-5 to -30°C). | |
| Genome and DNA Sequencing | |
| Targeted Sequencing Library | |
| Dry Ice |
| DNA | |
| Ion PGM™ System | |
| Oncomine | |
| DNA Extraction |