missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Omnifit™ EZ Benchmark Columns with 2 Fixed Endpieces

Catalog No. 111009256
Change view
Click to view available options
Length (Metric):
250 mm
330 mm
400 mm
500 mm
Diameter (Metric):
6.6 mm
10 mm
15 mm
25 mm
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
111009256 250 mm 6.6 mm
111009126 400 mm 6.6 mm
111009087 330 mm 10 mm
111009088 400 mm 10 mm
111008959 500 mm 10 mm
111008843 330 mm 15 mm
111008794 400 mm 15 mm
111008747 500 mm 15 mm
111008620 330 mm 25 mm
111008633 400 mm 25 mm
111008550 500 mm 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 111009256 Supplier MilliporeSigma™ Supelco™ Supplier No. K5128U

Omnifit™ Labware complete columns are supplied assembled and ready for use, with tubing and fittings to connect the column into an existing system.

This configuration of Benchmark columns is non-adjustable with two fixed endpieces (-FF), a cost-effective option when height adjustment is not important.

Specifications

Diameter (Metric) 6.6 mm
Length (Metric) 250 mm
Product Line Omnifit
Column Type Chromatography

Omnifit is a registered trademark of Diba Industries