missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Omeprazole, 98+%

$82.77 - $634.09

Chemical Identifiers

CAS 73590-58-6
Molecular Formula C17H19N3O3S
Molecular Weight (g/mol) 345.42
InChI Key SUBDBMMJDZJVOS-UHFFFAOYSA-N
Synonym omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac
PubChem CID 4594
ChEBI CHEBI:77260
IUPAC Name 6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
SMILES CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC458520010
View Documents Promotion Details
Thermo Scientific™
458520010
1g
Each for $82.77
Only null left
 
AC458520050
View Documents Promotion Details
Thermo Scientific™
458520050
5g
Each for $205.12
Only null left
 
AC458520250
View Documents Promotion Details
Thermo Scientific™
458520250
25g
Each for $634.09
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

73590-58-6
345.42
omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac
CHEBI:77260
CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC
C17H19N3O3S
SUBDBMMJDZJVOS-UHFFFAOYSA-N
4594
6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole

Specifications

73590-58-6
Authentic
Glass Bottle
omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac
SUBDBMMJDZJVOS-UHFFFAOYSA-N
6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
4594
345.42
Omeprazole
0.5% max. (60°C, 4 h) (vacuum)
98% min. (HPLC)
C17H19N3O3S
Solubility in water: insoluble in water. Other solubilities: very soluble in chloroform, dimethylformamide, slightly soluble in methanol,ethanol, very slighly soluble in acetonitrile, isopropanol
CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC
345.42
CHEBI:77260
98+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only