missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Omeprazole, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 158.0°C to 159.0°C |
| Loss on Drying | 0.5% max. (60°C, 4 h) (vacuum) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% min. (HPLC) |
| Quantity | 5 g |
| Solubility Information | Solubility in water: insoluble in water. Other solubilities: very soluble in chloroform, dimethylformamide, slightly soluble in methanol,ethanol, very slighly soluble in acetonitrile, isopropanol |
| Formula Weight | 345.42 |
| Percent Purity | 98+% |
| Chemical Name or Material | Omeprazole |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and kee |
RUO – Research Use Only