missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Omeprazole, 98+%

Supplier:  Thermo Scientific™ 458520010

 View more versions of this product

Catalog No. AC458520010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Omeprazole
0.5% max. (60°C, 4 h) (vacuum)
98% min. (HPLC)
C17H19N3O3S
omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac
SUBDBMMJDZJVOS-UHFFFAOYSA-N
6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
4594
345.42
73590-58-6
Authentic
Glass Bottle
1g
Solubility in water: insoluble in water. Other solubilities: very soluble in chloroform, dimethylformamide, slightly soluble in methanol,ethanol, very slighly soluble in acetonitrile, isopropanol
CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC
345.42
CHEBI:77260
98+%

Chemical Identifiers

73590-58-6
345.42
omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac
CHEBI:77260
CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC
C17H19N3O3S
SUBDBMMJDZJVOS-UHFFFAOYSA-N
4594
6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only