missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Olopatadine hydrochloride

Catalog No. p-7214741
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AC467710010 1 g
AC467710050 5 g
2 options

Catalog No. AC467710010

Supplier: Thermo Scientific Chemicals 467710010

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 140462-76-6
Molecular Formula C21H24ClNO3

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: soluble.
Melting Point 248.0°C
Formula Weight 373.88
InChI Key HVRLZEKDTUEKQH-NOILCQHBSA-N
SMILES [H+].[Cl-].CN(C)CC\C=C1\C2=CC=CC=C2COC2=CC=C(CC(O)=O)C=C12
IUPAC Name hydrogen 2-[(2Z)-2-[3-(dimethylamino)propylidene]-9-oxatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-5-yl]acetic acid chloride
Molecular Weight (g/mol) 373.88
Chemical Name or Material Olopatadine hydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Very toxic to aquatic life.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

RUO – Research Use Only