missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Olopatadine hydrochloride

Catalog No. AC467710010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467710010 1 g
AC467710050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467710010 Supplier Thermo Scientific Chemicals Supplier No. 467710010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 140462-76-6
Molecular Formula C21H24ClNO3

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: soluble.
Melting Point 248.0°C
Formula Weight 373.88
InChI Key HVRLZEKDTUEKQH-NOILCQHBSA-N
SMILES [H+].[Cl-].CN(C)CC\C=C1\C2=CC=CC=C2COC2=CC=C(CC(O)=O)C=C12
IUPAC Name hydrogen 2-[(2Z)-2-[3-(dimethylamino)propylidene]-9-oxatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-5-yl]acetic acid chloride
Molecular Weight (g/mol) 373.88
Chemical Name or Material Olopatadine hydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Very toxic to aquatic life.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

RUO – Research Use Only