missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Olopatadine hydrochloride

Catalog No. 424150
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
424150 50 mg
424110 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 424150 Supplier R&D Systems™ Supplier No. 4241/50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. H1 antagonist; ocular antiallergic agent

Histamine H1 receptor antagonist (Ki = 31.6 nM). Inhibits the release of histamine, prostaglandin D2 and tryptase in a concentration-dependent manner. Mast cell stabilizer; inhibits mast cell mediator release. Also suppresses inflammation by inhibition of cytokine production.

Chemical Identifiers

CAS 140462-76-6
Molecular Formula C21H24ClNO3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 373.87
Purity >99%
Molecular Weight (g/mol) 373.877
Chemical Name or Material Olopatadine hydrochloride
Recommended Storage Desiccate at Room Temperature