missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ OLIGO™ R3 Reversed Phase Columns

Catalog No. p-200156827
Change view
Click to view available options
Dimensions:
4.6 x 100 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions
1133226 4.6 x 100 mm
1133426 4.6 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1133226 Supplier Thermo Scientific™ Supplier No. 1133226
Only null left

Thermo Scientific™ Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules.

Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules. The combination of 30-μm particle size, resistance to high pH, and high-phase ratio, makes Oligo R3 resin particularly suited to preparative purification of synthetic oligonucleotides and peptides. The packing consists of cross-linked poly-divinylbenzene particles.

Oligo R3 bulk resin differs somewhat in selectivity from conventional silica-based, reversed-phase resins and is similar to high-carbon-loading through C18 supports. In addition, the complete lack of residual silanol activity may modify the selectivity of ionic species.

Features of Oligo R3 resin and pre-packed columns include:

  • High performance chromatography resin for analytical to process scale separations
  • High resolution preparative purification at high flow rates
  • Chemical stability—allows aggressive cleaning and sanitization
  • Poly-divinylbenzene particles—yield a robust, easily packable matrix
  • Combines low back pressure, high pH stability, and high capacity

Specifications

Column Format Prepacked Column
Diameter (Metric) 4.6 mm
Length (Metric) 100 mm
Max. Pressure 2400 psi (170 bar)
Particle Size 30 μm
Pore Size 350 Å
Quantity 1 Column
Stationary Phase Phenyl
Content And Storage Store at room temperature.
Product Line POROS
Shipping Condition Room Temperature
Volume (Metric) 1.7 mL
Dimensions 4.6 x 100 mm
Column Type Reversed Phase
Material PEEK
Type Analytical Column
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.