missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleoyl-L-carnitine, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
$259.19 - $259.19
Specifications
| CAS | 38677-66-6 |
|---|---|
| Linear Formula | C25H47 NO4 |
| MDL Number | MFCD22416941 |
| Synonym | (2 R)-3-Carboxy-N,N,N-trimethyl-2-[[(9 Z)-1-oxo-9-octadecen-1-yl]oxy]-1-propanaminium inner salt; cis-9-Octadecenoyl-L-carnitine |
| Type | Analytical Standard |
Specifications
| 38677-66-6 | |
| MFCD22416941 | |
| Analytical Standard | |
| ≥95.0% (HPLC) |
| C25H47 NO4 | |
| (2 R)-3-Carboxy-N,N,N-trimethyl-2-[[(9 Z)-1-oxo-9-octadecen-1-yl]oxy]-1-propanaminium inner salt; cis-9-Octadecenoyl-L-carnitine | |
| 425.64 | |
| Analytical Standard |