missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleoyl-L-carnitine, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 38677-66-6 |
| Linear Formula | C25H47 NO4 |
| MDL Number | MFCD22416941 |
| Synonym | (2 R)-3-Carboxy-N,N,N-trimethyl-2-[[(9 Z)-1-oxo-9-octadecen-1-yl]oxy]-1-propanaminium inner salt; cis-9-Octadecenoyl-L-carnitine |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 425.64 |
| Percent Purity | ≥95.0% (HPLC) |
| Grade | Analytical Standard |