missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Olaparib

Catalog No. 757950
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
757950 50 mg
757910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 757950 Supplier Tocris Bioscience™ Supplier No. 7579/50

Potent PARP inhibitor

Olaparib is a potent PARP2 and PARP1 inhibitor (IC50 values are 1 and 5 nM, respectively). Olaparib abolishes PARP-1 activity at a concentration of 30-100 nM in SW620 cell lysates. BRCA1-deficient cell lines are hypersensitive to PARP inhibition by Olaparib in comparison with BRCA1- and BRCA2-proficient cells. Olaparib potentiates the antitumor activity of the methylating chemotherapeutic agent Temozolomide (Cat. No. 2706) in xenograft mouse model. Olaparib is orally bioavailable in rats.

Specifications

Chemical Name or Material 4-[[3-[[4-(Cyclopropylcarbonyl)-1-piperazinyl]carbonyl]-4-fluorophenyl]methyl]-1(2 H)-phthalazinone
CAS 763113-22-0
Quantity 50 mg
Target Poly(ADP-ribose) Polymerase Inhibitors
Molecular Formula C24H23FN4O3
Purity 0.98