missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ School Balance

Catalog No. S63485
Change view
Click to view available options
Weight (Metric):
0.46 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
S63485 0.46 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog No. S63485 Supplier OHAUS™ Supplier No. SB1200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Ideal teaching tool to introduce students to precision mass measurement. OHAUS School Balances measure small items such as paper clips, marbles and coins, as well as heavier objects such as books.

  • Dual pointers allow students to view results from either side of the balance
  • Interchangeable square pans remove for easy pouring, cleaning and storing
  • Pivoting beam provides accuracy to 0.5g
  • Capacity: 70.5 oz. (2000g)
  • Includes 18-piece plastic weight set

School Balance Measurement Bundle

  • Includes six Ohaus school balances (S01976) and one measurement skills assessment kit (S97292)

Specifications

Depth (English) 4 in.
Depth (English) Pan 1.25 in.
Depth (Metric) 10 cm
Depth (Metric) Pan 3.2 cm
Height (English) 4.5 in.
Height (Metric) 11.5 cm
Includes 8 Piece Metal mass set, bowl type pans for use in
Material ABS Plastic
Weight (English) 1.01 lb.
Width (English) 11 in.
Weight (Metric) 0.46 kg
Width (Metric) 28 cm
Warranty 1 Year
Capacity 4.4 lb., 2 kg
Display Type Analog
Type School Balance
Show More Show Less