missing translation for 'onlineSavingsMsg'
Learn More

Octaphenylcyclotetrasiloxane 98.0+%, TCI America™

Supplier:  TCI America O023125G

 View more versions of this product

Catalog No. O023125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Octaphenylcyclotetrasiloxane
201°C
332°C
MFCD00003268
octaphenylcyclotetrasiloxane, cyclotetrasiloxane, octaphenyl, 2,2,4,4,6,6,8,8-octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, octaphenyltetracyclosiloxane, cyclotetrasiloxane, 2,2,4,4,6,6,8,8-octaphenyl, 1,1,3,3,5,5,7,7-octaphenylcyclotetrasiloxane, octaphenyl tetracyclosiloxane, acmc-1aqwg, ph2sio 4, vsikjpjinidelz-uhfffaoysa
O1[Si](O[Si](O[Si](O[Si]1(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
793.18
793.18
Crystalline Powder
546-56-5
White
C48H40O4Si4
25 g
VSIKJPJINIDELZ-UHFFFAOYSA-N
octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
68347
≥98.0% (HPLC)

Chemical Identifiers

546-56-5
793.18
VSIKJPJINIDELZ-UHFFFAOYSA-N
68347
O1[Si](O[Si](O[Si](O[Si]1(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C48H40O4Si4
MFCD00003268
octaphenylcyclotetrasiloxane, cyclotetrasiloxane, octaphenyl, 2,2,4,4,6,6,8,8-octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, octaphenyltetracyclosiloxane, cyclotetrasiloxane, 2,2,4,4,6,6,8,8-octaphenyl, 1,1,3,3,5,5,7,7-octaphenylcyclotetrasiloxane, octaphenyl tetracyclosiloxane, acmc-1aqwg, ph2sio 4, vsikjpjinidelz-uhfffaoysa
octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : GZ4398500

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions