missing translation for 'onlineSavingsMsg'
Learn More

Octaphenylcyclotetrasiloxane 98.0+%, TCI America™

Catalog No. p-7134250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02315G 5 g
O023125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02315G Supplier TCI America Supplier No. O02315G
Only null left

Chemical Identifiers

CAS 546-56-5
Molecular Formula C48H40O4Si4
Molecular Weight (g/mol) 793.18
MDL Number MFCD00003268
InChI Key VSIKJPJINIDELZ-UHFFFAOYSA-N
Synonym octaphenylcyclotetrasiloxane, cyclotetrasiloxane, octaphenyl, 2,2,4,4,6,6,8,8-octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, octaphenyltetracyclosiloxane, cyclotetrasiloxane, 2,2,4,4,6,6,8,8-octaphenyl, 1,1,3,3,5,5,7,7-octaphenylcyclotetrasiloxane, octaphenyl tetracyclosiloxane, acmc-1aqwg, ph2sio 4, vsikjpjinidelz-uhfffaoysa
PubChem CID 68347
IUPAC Name octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
SMILES O1[Si](O[Si](O[Si](O[Si]1(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 201°C
Color White
Boiling Point 332°C
Quantity 5 g
Formula Weight 793.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Octaphenylcyclotetrasiloxane
RTECSNumber GZ4398500
TSCA Yes