accessibility menu, dialog, popup

UserName

OSiOSiOSiOSi

CAS RN 546-56-5

IUPAC Name: octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Synonyms: octaphenyl tetracyclosiloxane
Molecular Weight (g/mol): 793.18
Molecular Formula: C48H40O4Si4
InChi Key: VSIKJPJINIDELZ-UHFFFAOYSA-N
SMILES: O1[Si](O[Si](O[Si](O[Si]1(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)