missing translation for 'onlineSavingsMsg'
Learn More

Octanoyl chloride, 99%

CAS: 111-64-8 | C8H15ClO | 162.657 g/mol

Supplier:  Thermo Scientific Chemicals L03526AP

 View more versions of this product

Catalog No. AAL03526AP

Only null left
Explore available promotions

Description

Description

Octanoyl chloride is used to produce adhesives. It is used as an acylating agent for a variety of compounds such as sugars (e.g. Sucrose, aromatic compounds (e.g. Anisole and monoglycerides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanoyl chloride is used to produce adhesives. It is used as an acylating agent for a variety of compounds such as sugars (e.g. Sucrose, aromatic compounds (e.g. Anisole and monoglycerides.

Solubility
Soluble in organic solvents (acetone, chloroform, toluene, THF). Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with Bases, Alcohols, Amines, Oxidizing agents. Store away from alcohol and amines.
Specifications

Specifications

Octanoyl chloride
-63°C
195°C
Pungent
1.435
500 mL
635917
capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride
REEZZSHJLXOIHL-UHFFFAOYSA-N
octanoyl chloride
8124
99%
111-64-8
0.942
80°C (176°F)
C8H15ClO
MFCD00000766
UN3265
Moisture sensitive
Soluble in organic solvents (acetone,chloroform,toluene,THF). Hydrolyzes in water.
CCCCCCCC(=O)Cl
162.657
162.66

Chemical Identifiers

111-64-8
162.657
REEZZSHJLXOIHL-UHFFFAOYSA-N
8124
CCCCCCCC(=O)Cl
C8H15ClO
MFCD00000766
capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride
octanoyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.

P210-P233-P234-P240-P241-P242-P243-P260-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P310-P333+P313-P362-P374-P390-P501c

H227-H290-H315-H317-H318-H330

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 203-891-6

RTECSNumber : RH1570000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only