missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octanoyl chloride, 99%
Description
Octanoyl chloride is used to produce adhesives. It is used as an acylating agent for a variety of compounds such as sugars (e.g. Sucrose, aromatic compounds (e.g. Anisole and monoglycerides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Octanoyl chloride is used to produce adhesives. It is used as an acylating agent for a variety of compounds such as sugars (e.g. Sucrose, aromatic compounds (e.g. Anisole and monoglycerides.
Solubility
Soluble in organic solvents (acetone, chloroform, toluene, THF). Hydrolyzes in water.
Notes
Moisture Sensitive. Incompatible with Bases, Alcohols, Amines, Oxidizing agents. Store away from alcohol and amines.
Octanoyl chloride is used to produce adhesives. It is used as an acylating agent for a variety of compounds such as sugars (e.g. Sucrose, aromatic compounds (e.g. Anisole and monoglycerides.
Solubility
Soluble in organic solvents (acetone, chloroform, toluene, THF). Hydrolyzes in water.
Notes
Moisture Sensitive. Incompatible with Bases, Alcohols, Amines, Oxidizing agents. Store away from alcohol and amines.
Specifications
Specifications
| Melting Point | -63°C |
| Density | 0.942 |
| Boiling Point | 195°C |
| Flash Point | 80°C (176°F) |
| Odor | Pungent |
| Refractive Index | 1.435 |
| Quantity | 100 mL |
| UN Number | UN3265 |
| Beilstein | 635917 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H227-H290-H315-H317-H318-H330 |
| Hazard Statement | GHS H Statement H314-H318-H302-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Combustible liquid. |
| Precautionary Statement | P210-P233-P234-P240-P241-P242-P243-P260-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P310-P333+P313-P362-P374-P390-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: II |
| EINECSNumber | 203-891-6 |
| RTECSNumber | RH1570000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only