accessibility menu, dialog, popup

UserName

H3CClO

CAS RN 111-64-8

IUPAC Name: octanoyl chloride
Synonyms: octanoyl chloride
Molecular Weight (g/mol): 162.66
Molecular Formula: C8H15ClO
InChi Key: REEZZSHJLXOIHL-UHFFFAOYSA-N
SMILES: CCCCCCCC(Cl)=O
Boiling Point 
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)