missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octafluoronaphthalene 92.0+%, TCI America™
Supplier: TCI America O01585G
Specifications
| Octafluoronaphthalene | |
| 87°C | |
| 209°C | |
| MFCD00014307 | |
| octafluoronaphthalene, perfluoronaphthalene, naphthalene, octafluoro, octafluoronapthalene, octafiuoronaphthalene, acmc-1cr7y, octafluoronaphthalene 1g, octafluoronaphthalene radical anion | |
| C12=C(C(=C(C(=C1F)F)F)F)C(=C(C(=C2F)F)F)F | |
| 272.097 | |
| 272.10 | |
| Crystalline Powder |
| 313-72-4 | |
| White-Yellow | |
| C10F8 | |
| 5 g | |
| JDCMOHAFGDQQJX-UHFFFAOYSA-N | |
| 1,2,3,4,5,6,7,8-octafluoronaphthalene | |
| 67564 | |
| ≥92.0% (GC) |
Chemical Identifiers
| 313-72-4 | |
| 272.097 | |
| JDCMOHAFGDQQJX-UHFFFAOYSA-N | |
| 67564 | |
| C12=C(C(=C(C(=C1F)F)F)F)C(=C(C(=C2F)F)F)F |
| C10F8 | |
| MFCD00014307 | |
| octafluoronaphthalene, perfluoronaphthalene, naphthalene, octafluoro, octafluoronapthalene, octafiuoronaphthalene, acmc-1cr7y, octafluoronaphthalene 1g, octafluoronaphthalene radical anion | |
| 1,2,3,4,5,6,7,8-octafluoronaphthalene |
Safety and Handling
TSCA : No