accessibility menu, dialog, popup

UserName

CAS RN 313-72-4

IUPAC Name: octafluoronaphthalene
Synonyms: octafluoronaphthalene
Molecular Weight (g/mol): 272.1
Molecular Formula: C10F8
InChi Key: JDCMOHAFGDQQJX-UHFFFAOYSA-N
SMILES: FC1=C(F)C2=C(F)C(F)=C(F)C(F)=C2C(F)=C1F
Boiling Point 
  • (5)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (5)
  • (1)
  • (1)
  • (5)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)