missing translation for 'onlineSavingsMsg'
Learn More

Octafluoronaphthalene 92.0+%, TCI America™

Catalog No. p-7134212
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01581G 1 g
O01585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01581G Supplier TCI America Supplier No. O01581G
Only null left

Chemical Identifiers

CAS 313-72-4
Molecular Formula C10F8
Molecular Weight (g/mol) 272.097
MDL Number MFCD00014307
InChI Key JDCMOHAFGDQQJX-UHFFFAOYSA-N
Synonym octafluoronaphthalene, perfluoronaphthalene, naphthalene, octafluoro, octafluoronapthalene, octafiuoronaphthalene, acmc-1cr7y, octafluoronaphthalene 1g, octafluoronaphthalene radical anion
PubChem CID 67564
IUPAC Name 1,2,3,4,5,6,7,8-octafluoronaphthalene
SMILES C12=C(C(=C(C(=C1F)F)F)F)C(=C(C(=C2F)F)F)F

Specifications

Melting Point 87°C
Color White-Yellow
Boiling Point 209°C
Quantity 1 g
Formula Weight 272.10
Percent Purity ≥92.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Octafluoronaphthalene