missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ O8 OGG1 Inhibitor

Catalog No. 623625
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
623625 25 mg
62365 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 623625 Supplier Tocris Bioscience™ Supplier No. 6236/25

Potent and selective 8-oxoguanine DNA glycosylase 1 (OGG1) inhibitor (IC50 = 0.2 μM in a gel based assay)

O8 OGG1 Inhibitor is a potent and selective 8-oxoguanine DNA glycosylase 1 (OGG1) inhibitor (IC50 = 0.2 μM in a gel based assay). Exhibits > 200-fold selectivity for OGG1 over other DNA glycosylases, NEIL1 and NTH1.

Specifications

Chemical Name or Material 3,4-Dichlorobenzo[b]thiophene-2-carbohydrazide
CAS 350997-39-6
Quantity 25 mg
Molecular Formula C9H6Cl2N2OS
Formula Weight 261.13
Purity 98%