missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Xylyl Cyanide 98.0+%, TCI America™
Supplier: TCI America X0045100ML
Specifications
| o-Xylyl Cyanide | |
| Yellow | |
| C9H9N | |
| 100 mL | |
| 2-methylbenzyl cyanide, 2-o-tolyl acetonitrile, o-tolylacetonitrile, 2-methylphenylacetonitrile, o-methylphenylacetonitrile, o-xylyl cyanide, o-methylbenzyl cyanide, acetonitrile, o-tolyl, benzeneacetonitrile, 2-methyl, 2-2-methylphenyl acetonitrile | |
| CC1=CC=CC=C1CC#N | |
| 131.178 | |
| CHEBI:27982 | |
| ≥98.0% (GC) |
| 22364-68-7 | |
| 120°C | |
| MFCD00001904 | |
| 3276 | |
| WMGVPDQNPUQRND-UHFFFAOYSA-N | |
| 2-(2-methylphenyl)acetonitrile | |
| 31155 | |
| 131.18 | |
| Liquid |
Chemical Identifiers
| 22364-68-7 | |
| 131.178 | |
| WMGVPDQNPUQRND-UHFFFAOYSA-N | |
| 31155 | |
| 2-(2-methylphenyl)acetonitrile |
| C9H9N | |
| MFCD00001904 | |
| 2-methylbenzyl cyanide, 2-o-tolyl acetonitrile, o-tolylacetonitrile, 2-methylphenylacetonitrile, o-methylphenylacetonitrile, o-xylyl cyanide, o-methylbenzyl cyanide, acetonitrile, o-tolyl, benzeneacetonitrile, 2-methyl, 2-2-methylphenyl acetonitrile | |
| CHEBI:27982 | |
| CC1=CC=CC=C1CC#N |
Safety and Handling
RTECSNumber : AM2290000
TSCA : No