missing translation for 'onlineSavingsMsg'
Learn More

o-Xylyl Cyanide 98.0+%, TCI America™

Catalog No. p-7137962
Change view
Click to view available options
Quantity:
25 mL
100 mL
Catalog No. Quantity
X004525ML 25 mL
X0045100ML 100 mL
2 options

Catalog No. X004525ML

Supplier: TCI America X004525ML

Only null left

Chemical Identifiers

CAS 22364-68-7
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00001904
InChI Key WMGVPDQNPUQRND-UHFFFAOYSA-N
Synonym 2-methylbenzyl cyanide, 2-o-tolyl acetonitrile, o-tolylacetonitrile, 2-methylphenylacetonitrile, o-methylphenylacetonitrile, o-xylyl cyanide, o-methylbenzyl cyanide, acetonitrile, o-tolyl, benzeneacetonitrile, 2-methyl, 2-2-methylphenyl acetonitrile
PubChem CID 31155
ChEBI CHEBI:27982
IUPAC Name 2-(2-methylphenyl)acetonitrile
SMILES CC1=CC=CC=C1CC#N

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 mL
UN Number 3276
Formula Weight 131.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material o-Xylyl Cyanide
RTECSNumber AM2290000
TSCA No