missing translation for 'onlineSavingsMsg'
Learn More

o-Xylene 98.0+%, TCI America™

Catalog No. p-7137952
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
X001225ML 25 mL
X0012500ML 500 mL
2 options

Catalog No. X001225ML

Supplier: TCI America X001225ML

Only null left

Chemical Identifiers

CAS 95-47-6
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
PubChem CID 7237
ChEBI CHEBI:28063
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

Melting Point -23°C
Color Colorless
Boiling Point 145°C
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 1307
Formula Weight 106.17
Percent Purity ≥98.0% (GC)
Chemical Name or Material o-Xylene
EINECSNumber (3)-0003&(3)-0060
RTECSNumber ZE2450000
TSCA Yes