missing translation for 'onlineSavingsMsg'
Learn More

o-Xylene, HPLC Grade, 96% min

Catalog No. AA22902K2
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA22902K2 1 L
AA22902M1 4 x 1 L
AA22902K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. AA22902K2 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 022902K2

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.04% | CAS: 95-47-6 | C8H10 | 106.17 g/mol

o-Xylene is largely used in the production of phthalic anhydride, and is generally extracted by distillation from a mixed Xylene stream in a plant primarily designed for p-Xylene production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-47-6
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
PubChem CID 7237
ChEBI CHEBI:28063
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

Melting Point -25°C
Density 0.877
Quantity 1 L
Flash Point 32°C (89°F)
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.505
Specific Gravity 0.877
Linear Formula (CH3)2C6H4
UN Number UN1307
Beilstein 1815558
Boiling Point 143°C to 145°C
Solubility Information Soluble in ethanol,diethyl ether,water(0.2g/L).
Formula Weight 106.17
Percent Purity ≥96%
Grade HPLC
Chemical Name or Material o-Xylene
Show More Show Less
missing translation for 'healthHazard1' H226-H304-H312+H332-H315-H319-H335-H336-H373
missing translation for 'healthHazard3' P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 202-422-2
RTECSNumber ZE2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only