missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Xylene, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 9566050ML
Specifications
| 95-47-6 | |
| 3.7 (vs. air) | |
| Neat | |
| MFCD00008519 | |
| 143°C to 145°C | |
| 50 mL | |
| ≥99.0% (GC) |
| 16 mmHg (37.7°C) | |
| 0.879 g/mL (at 20°C) | |
| C6H4(CH3)2 | |
| -26°C to +23°C | |
| 1,2-Dimethylbenzene | |
| 106.17 | |
| Analytical Standard |