missing translation for 'onlineSavingsMsg'
Learn More

o-Vanillin 99.0+%, TCI America™

Supplier:  TCI America H0262500G

 View more versions of this product

Catalog No. H0262500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

o-Vanillin
44°C
C8H8O3
500 g
JJVNINGBHGBWJH-UHFFFAOYSA-N
2-hydroxy-3-methoxybenzaldehyde
8991
152.15
Crystalline Powder
148-53-8
268°C
MFCD00003322
3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehyde
COC1=CC=CC(C=O)=C1O
152.15
CHEBI:78339
≥99.0% (GC)

Chemical Identifiers

148-53-8
152.15
JJVNINGBHGBWJH-UHFFFAOYSA-N
8991
2-hydroxy-3-methoxybenzaldehyde
C8H8O3
MFCD00003322
3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehyde
CHEBI:78339
COC1=CC=CC(C=O)=C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1200

RTECSNumber : CU6530000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions