accessibility menu, dialog, popup

UserName

CH3OOHO

CAS RN 148-53-8

IUPAC Name: 2-hydroxy-3-methoxybenzaldehyde
Synonyms: O-vanillin ortho-vanillin
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: JJVNINGBHGBWJH-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C=O)=C1O
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (3)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)