missing translation for 'onlineSavingsMsg'
Learn More

o-Vanillin 99.0+%, TCI America™

Catalog No. p-7130448
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
H026225G 25 g
H0262100G 100 g
H0262500G 500 g
3 options

Catalog No. H026225G

Supplier: TCI America H026225G

Only null left

Chemical Identifiers

CAS 148-53-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00003322
InChI Key JJVNINGBHGBWJH-UHFFFAOYSA-N
Synonym 3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehyde
PubChem CID 8991
ChEBI CHEBI:78339
IUPAC Name 2-hydroxy-3-methoxybenzaldehyde
SMILES COC1=CC=CC(C=O)=C1O

Specifications

Melting Point 44°C
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material o-Vanillin
EINECSNumber (3)-1200
RTECSNumber CU6530000
TSCA Yes