missing translation for 'onlineSavingsMsg'
Learn More

o-Tolyl Acetate 98.0+%, TCI America™

Catalog No. A066925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A066925G 25 g
A0669500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A066925G Supplier TCI America Supplier No. A066925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 533-18-6
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00053704
InChI Key AMZORBZSQRUXNC-UHFFFAOYSA-N
Synonym Acetic Acid o-Cresyl Ester, Acetic Acid o-Tolyl Ester, o-Cresyl Acetate
PubChem CID 10778
IUPAC Name 2-methylphenyl acetate
SMILES CC(=O)OC1=CC=CC=C1C

Specifications

Color Colorless
Boiling Point 208°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material o-Tolyl Acetate
EINECSNumber (3)-0644
TSCA Yes