missing translation for 'onlineSavingsMsg'
Learn More

o-Toluidine-4-sulfonic Acid Sodium Salt Tetrahydrate 98.0+%, TCI America™

Catalog No. T030425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T030425G 25 g
T0304500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T030425G Supplier TCI America Supplier No. T030425G

Chemical Identifiers

CAS 63450-43-1
Molecular Formula C7H8NNaO3S
Molecular Weight (g/mol) 209.195
MDL Number MFCD00070549
InChI Key RVSNUNNCRYLEEM-UHFFFAOYSA-M
Synonym 2-Aminotoluene-5-sulfonic Acid Sodium Salt, Sodium 2-Aminotoluene-5-sulfonate, Sodium o-Toluidine-4-sulfonate, 4-Amino-3-methylbenzenesulfonic Acid Sodium Salt, Sodium 4-Amino-3-methylbenzenesulfonate
PubChem CID 23695986
IUPAC Name sodium;4-amino-3-methylbenzenesulfonate
SMILES CC1=C(C=CC(=C1)S(=O)(=O)[O-])N.[Na+]

Specifications

Quantity 25 g
Formula Weight 209.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material o-Toluidine-4-sulfonic Acid Sodium Salt Tetrahydrate
EINECSNumber (3)-1979
RTECSNumber DB4979000
TSCA No