CAS RN 63450-43-1
IUPAC Name:
sodium 4-amino-3-methylbenzene-1-sulfonate
Synonyms:
sodium 4-amino-3-methylbenzenesulfonate
Molecular Weight (g/mol):
209.19
Molecular Formula:
C7H8NNaO3S
InChi Key:
RVSNUNNCRYLEEM-UHFFFAOYSA-M
SMILES:
[Na+].CC1=CC(=CC=C1N)S([O-])(=O)=O
1
–
1
of
1
results
1
o-Toluidine-4-sulfonic Acid Sodium Salt Tetrahydrate 98.0+%, TCI America™
CAS: 63450-43-1 Molecular Formula: C7H8NNaO3S Molecular Weight (g/mol): 209.195 MDL Number: MFCD00070549 InChI Key: RVSNUNNCRYLEEM-UHFFFAOYSA-M Synonym: 2-Aminotoluene-5-sulfonic Acid Sodium Salt, Sodium 2-Aminotoluene-5-sulfonate, Sodium o-Toluidine-4-sulfonate, 4-Amino-3-methylbenzenesulfonic Acid Sodium Salt, Sodium 4-Amino-3-methylbenzenesulfonate PubChem CID: 23695986 IUPAC Name: sodium;4-amino-3-methylbenzenesulfonate SMILES: CC1=C(C=CC(=C1)S(=O)(=O)[O-])N.[Na+]