missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Toluenethiol, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 14.0°C |
| Color | Colorless to Yellow |
| Density | 1.0540g/mL |
| Boiling Point | 194.0°C |
| Flash Point | 63°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5790 to 1.5810 |
| Linear Formula | CH3C6H4SH |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye damage. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF SWALLOWED: rinse mouth. Do |
| EINECSNumber | 205-276-8 |
| RTECSNumber | XT8750000 |
| TSCA | TSCA |
RUO – Research Use Only