missing translation for 'onlineSavingsMsg'
Learn More

o-Toluenethiol, 97%

Catalog No. AC259240050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC259240050 5 g Glass bottle
AC259240250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC259240050 Supplier Thermo Scientific Chemicals Supplier No. 259240050

CAS: 137-06-4 | C7H8S | 124.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 137-06-4
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004838
InChI Key LXUNZSDDXMPKLP-UHFFFAOYSA-N
Synonym o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl
PubChem CID 8712
IUPAC Name 2-methylbenzenethiol
SMILES CC1=CC=CC=C1S

Specifications

Melting Point 14.0°C
Color Colorless to Yellow
Density 1.0540g/mL
Boiling Point 194.0°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5790 to 1.5810
Linear Formula CH3C6H4SH
Quantity 5 g
Beilstein 06,37
Merck Index 15,9478
Specific Gravity 1.054
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 124.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material o-Toluenethiol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED: rinse mouth.
Do
EINECSNumber 205-276-8
RTECSNumber XT8750000
TSCA TSCA

RUO – Research Use Only