missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine, Powder, Reagent, Spectrum™ Chemical

Catalog No. p-7157917
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604912 25 g Amber Glass Bottle
18604913 100 g Amber Glass Bottle
18604914 500 g Amber Glass Bottle
18604915 2.5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604912 Supplier Spectrum Chemical Mfg Cor Supplier No. T108425GM
Only null left

T1084, 119-93-7, C14H16N2

Spectrum™ Chemical o-Tolidine, Powder, Reagent is a commercially important aromatic amine used to make dyes. It is also used in the production of certain elastomers and as a reagent and indicator in analytical forensic and clinical chemistry. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 119-93-7
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.30
InChI Key NUIURNJTPRWVAP-UHFFFAOYSA-N
IUPAC Name 3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diamine
SMILES CC1=CC(=CC=C1N)C1=CC=C(N)C(C)=C1

Specifications

CAS Max % 1
Melting Range 125-132°C
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 97.5%
Grade Reagent
Physical Form Crystalline Powder or Lumps