Learn More
o-Tolidine dihydrochloride, 99%
CAS: 612-82-8 | C14H16N2·2HCl | 285.2 g/mol
Supplier: Thermo Scientific Chemicals 421131000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| o-Tolidine dihydrochloride | |
| 130.0°C | |
| Authentic | |
| Glass Bottle | |
| 100 g | |
| 3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl | |
| CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl | |
| 285.2 | |
| 285.2 | |
| Crystalline Powder |
| 612-82-8 | |
| Beige to Beige-Gray | |
| 98.5% min. (on dry substance) (Argentometry) | |
| C14H16N2·2HCl | |
| 15,9675 | |
| LUKPNZHXJRJBAN-UHFFFAOYSA-N | |
| 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride | |
| 108938 | |
| 99% |
Chemical Identifiers
| 612-82-8 | |
| 285.2 | |
| 3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl | |
| 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride |
| C14H16N2·2HCl | |
| LUKPNZHXJRJBAN-UHFFFAOYSA-N | |
| 108938 | |
| CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl |
Safety and Handling
GHS H Statement
May cause cancer.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Danger
EINECSNumber : 210-322-5
RUO – Research Use Only