missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Tolidine, Certified, 0.1%, LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 91.37, 8.5, 0.13 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Linear Formula | C14H16N2·2HCl |
| UN Number | UN1789 |
| Solubility Information | Soluble in water |
| Formula Weight | 285.21 |
| Percent Purity | 0.1% |
| Grade | Certified |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May cause cancer. |
| Precautionary Statement | GHS P Statement Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. Take off contaminated clothing. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| Recommended Storage | Room Temperature |