missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine, Certified, 0.1%, LabChem™

Catalog No. LC261602
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC261602 1 L Poly Bottle
LC261601 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC261602 Supplier LabChem Supplier No. LC261602

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7732-18-5 | 285.21 g/mol

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0 , 612-82-8
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

Color Colorless
CAS Max % 91.37, 8.5, 0.13
Physical Form Liquid
Quantity 1 L
Linear Formula C14H16N2·2HCl
UN Number UN1789
Solubility Information Soluble in water
Formula Weight 285.21
Percent Purity 0.1%
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride; Carbon dioxide; Carbon monoxide; Nitrogen oxides
Chemical Name or Material o-Tolidine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause cancer.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature