missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine, 98%

CAS: 119-93-7 | C14H16N2 | 212.296 g/mol

Supplier:  Thermo Scientific Chemicals A1783722

 View more versions of this product

Catalog No. AAA1783722


Only null left
Explore available promotions

Description

Description

O-Tolidine is widely used in dye production. It is particularly used in textile, leather and paper industries. It can be used as an indicator in analytical, clinical and forensic chemistry. It is utilized for the determination of gold and chlorine level in swimming pool water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Tolidine is widely used in dye production. It is particularly used in textile, leather and paper industries. It can be used as an indicator in analytical, clinical and forensic chemistry. It is utilized for the determination of gold and chlorine level in swimming pool water.

Solubility
Slightly soluble in water.

Notes
Light and air sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

o-Tolidine
128°C to 132°C
244°C (471°F)
MFCD00014773
UN3077
14,9514
Slightly soluble in water.
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N
212.296
CHEBI:34320
98%
119-93-7
300°C to 301°C
C14H16N2
100 g
Light sensitive
o-tolidine, 3,3'-dimethylbenzidine, orthotolidine, diaminoditolyl, 2-tolidine, diaminotolyl, bianisidine, 3,3'-tolidine, 3,3'-dimethylbiphenyl-4,4'-diamine, 4,4'-bi-o-toluidine
NUIURNJTPRWVAP-UHFFFAOYSA-N
4-(4-amino-3-methylphenyl)-2-methylaniline
8413
212.3

Chemical Identifiers

119-93-7
212.296
NUIURNJTPRWVAP-UHFFFAOYSA-N
8413
4-(4-amino-3-methylphenyl)-2-methylaniline
C14H16N2
MFCD00014773
o-tolidine, 3,3'-dimethylbenzidine, orthotolidine, diaminoditolyl, 2-tolidine, diaminotolyl, bianisidine, 3,3'-tolidine, 3,3'-dimethylbiphenyl-4,4'-diamine, 4,4'-bi-o-toluidine
CHEBI:34320
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.

P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H350

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 204-358-0

RTECSNumber : DD1225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only